The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022574.4 | NP_072096.2 | 20 - 21 | 3609 | 71 |
XM_005250532.1 | XP_005250589.1 | 22 - 23 | 3390 | 71 | |
XM_011516472.1 | XP_011514774.1 | 22 - 23 | 3390 | 71 | |
XM_011516477.1 | XP_011514779.1 | 22 - 23 | 3387 | 71 | |
XM_017012526.1 | XP_016868015.1 | 22 - 23 | 3390 | 71 | |
XM_017012527.1 | XP_016868016.1 | 24 - 25 | 3288 | 71 | |
XM_017012528.1 | XP_016868017.1 | 23 - 24 | 2900 | 71 | |
XM_017012529.1 | XP_016868018.1 | 23 - 24 | 2907 | 71 | |
XM_017012530.1 | XP_016868019.1 | 23 - 24 | 3161 | 71 | |
XM_017012531.1 | XP_016868020.1 | 18 - 19 | 2413 | 71 | |
GenBank mRNA | AY176044.1 | - | 20 - 21 | 3810 | 71 |
BC018563.1 | - | 4 - 5 | 583 | 71 | |
BC129991.1 | - | 14 - 15 | 1757 | 71 |