The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022574.4 | NP_072096.2 | 7 - 8 | 1704 | 105 |
XM_005250532.1 | XP_005250589.1 | 9 - 10 | 1485 | 105 | |
XM_011516472.1 | XP_011514774.1 | 9 - 10 | 1485 | 105 | |
XM_011516477.1 | XP_011514779.1 | 9 - 10 | 1485 | 105 | |
XM_017012526.1 | XP_016868015.1 | 9 - 10 | 1485 | 105 | |
XM_017012527.1 | XP_016868016.1 | 11 - 12 | 1383 | 105 | |
XM_017012528.1 | XP_016868017.1 | 10 - 11 | 995 | 105 | |
XM_017012529.1 | XP_016868018.1 | 10 - 11 | 1002 | 105 | |
XM_017012530.1 | XP_016868019.1 | 10 - 11 | 1256 | 105 | |
XM_017012531.1 | XP_016868020.1 | 5 - 6 | 508 | 105 | |
GenBank mRNA | AY176044.1 | - | 7 - 8 | 1905 | 105 |