Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICA1
Entrez Gene ID
3382
Gene Name
islet cell autoantigen 1
Gene Aliases
ICA69, ICAp69
Chromosome Location
Chr.7: 8113184 - 8262687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136020.2 NP_001129492.1 12 - 13 1326 103
NM_001276478.1 NP_001263407.1 12 - 13 1308 103
NM_004968.3 NP_004959.2 12 - 13 1311 103
NM_022307.2 NP_071682.1 12 - 13 1126 103
XM_005249735.3 XP_005249792.1 12 - 13 1248 103
XM_011515345.1 XP_011513647.1 13 - 14 1413 103
XM_011515346.1 XP_011513648.1 13 - 14 1398 103
XM_011515347.2 XP_011513649.1 13 - 14 1285 103
XM_011515348.1 XP_011513650.1 13 - 14 1331 103
XM_011515349.1 XP_011513651.1 13 - 14 1264 103
XM_011515350.1 XP_011513652.1 13 - 14 1335 103
XM_011515351.1 XP_011513653.1 13 - 14 1470 103
XM_011515352.1 XP_011513654.1 13 - 14 1289 103
XM_011515353.2 XP_011513655.1 13 - 14 1342 103
XM_011515354.1 XP_011513656.1 10 - 11 926 103
XM_011515355.2 XP_011513657.1 8 - 9 819 103
XM_011515356.2 XP_011513658.1 8 - 9 773 103
XM_011515357.2 XP_011513659.1 8 - 9 627 103
XM_017012116.1 XP_016867605.1 13 - 14 1181 103
XM_017012117.1 XP_016867606.1 12 - 13 1284 103
XM_017012118.1 XP_016867607.1 12 - 13 1244 103
XM_017012119.1 XP_016867608.1 12 - 13 1219 103
XM_017012120.1 XP_016867609.1 12 - 13 1265 103
XM_017012121.1 XP_016867610.1 12 - 13 1132 103
XM_017012122.1 XP_016867611.1 13 - 14 1511 103
XM_017012123.1 XP_016867612.1 13 - 14 1450 103
XM_017012124.1 XP_016867613.1 13 - 14 1611 103
XM_017012125.1 XP_016867614.1 13 - 14 1361 103
GenBank mRNA AK292106.1 - 12 - 13 1195 103
BC008640.2 - 12 - 13 1243 103
EU416199.1 - 12 - 13 1146 103
L01100.1 - 12 - 13 1237 103
L21181.1 - 12 - 13 1135 103
U26591.1 - 12 - 13 1126 103
U26592.1 - 13 - 14 1397 103
U37183.1 - 10 - 11 1025 103

View Full Product Details