Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICA1
Entrez Gene ID
3382
Gene Name
islet cell autoantigen 1
Gene Aliases
ICA69, ICAp69
Chromosome Location
Chr.7: 8113184 - 8262687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136020.2 NP_001129492.1 2 - 3 286 141
NM_004968.3 NP_004959.2 2 - 3 271 141
XM_011515345.1 XP_011513647.1 2 - 3 286 141
XM_011515346.1 XP_011513648.1 2 - 3 271 141
XM_011515347.2 XP_011513649.1 2 - 3 158 141
XM_017012117.1 XP_016867606.1 2 - 3 244 141
XM_017012122.1 XP_016867611.1 2 - 3 257 141
XM_017012123.1 XP_016867612.1 2 - 3 196 141
GenBank mRNA AK292106.1 - 2 - 3 155 141
DA076379.1 - 2 - 3 272 141
DA154134.1 - 2 - 3 286 141
DB010744.1 - 2 - 3 255 141
DB040266.1 - 2 - 3 214 141
L21181.1 - 1 - 2 95 141
U26592.1 - 2 - 3 143 141
U38260.1 - 2 - 3 188 141

View Full Product Details