Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICA1
Entrez Gene ID
3382
Gene Name
islet cell autoantigen 1
Gene Aliases
ICA69, ICAp69
Chromosome Location
Chr.7: 8113184 - 8262687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136020.2 NP_001129492.1 8 - 9 1073 129
NM_001276478.1 NP_001263407.1 8 - 9 1055 129
NM_004968.3 NP_004959.2 8 - 9 1058 129
NM_022307.2 NP_071682.1 8 - 9 873 129
XM_005249735.3 XP_005249792.1 8 - 9 995 129
XM_011515345.1 XP_011513647.1 8 - 9 1073 129
XM_011515346.1 XP_011513648.1 8 - 9 1058 129
XM_011515347.2 XP_011513649.1 8 - 9 945 129
XM_011515348.1 XP_011513650.1 8 - 9 991 129
XM_011515349.1 XP_011513651.1 8 - 9 924 129
XM_011515350.1 XP_011513652.1 8 - 9 995 129
XM_011515351.1 XP_011513653.1 8 - 9 1130 129
XM_011515352.1 XP_011513654.1 8 - 9 949 129
XM_011515353.2 XP_011513655.1 8 - 9 1002 129
XM_011515354.1 XP_011513656.1 5 - 6 586 129
XM_011515355.2 XP_011513657.1 3 - 4 479 129
XM_011515356.2 XP_011513658.1 3 - 4 433 129
XM_011515357.2 XP_011513659.1 3 - 4 287 129
XM_017012116.1 XP_016867605.1 8 - 9 841 129
XM_017012117.1 XP_016867606.1 8 - 9 1031 129
XM_017012118.1 XP_016867607.1 8 - 9 991 129
XM_017012119.1 XP_016867608.1 8 - 9 966 129
XM_017012120.1 XP_016867609.1 8 - 9 1012 129
XM_017012121.1 XP_016867610.1 8 - 9 879 129
XM_017012122.1 XP_016867611.1 9 - 10 1258 129
XM_017012123.1 XP_016867612.1 9 - 10 1197 129
XM_017012124.1 XP_016867613.1 9 - 10 1358 129
XM_017012125.1 XP_016867614.1 9 - 10 1108 129
GenBank mRNA AK292106.1 - 8 - 9 942 129
BC008640.2 - 8 - 9 990 129
EU416199.1 - 7 - 8 806 129
L01100.1 - 8 - 9 984 129
L21181.1 - 8 - 9 882 129
U26591.1 - 8 - 9 873 129
U26592.1 - 9 - 10 1144 129
U37183.1 - 6 - 7 772 129

View Full Product Details