The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134758.3 | NP_001128230.1 | 2 - 3 | 242 | 82 |
NM_001267544.2 | NP_001254473.1 | 2 - 3 | 247 | 82 | |
NM_001267545.2 | NP_001254474.1 | 2 - 3 | 242 | 82 | |
NM_001267547.2 | NP_001254476.1 | 2 - 3 | 247 | 82 | |
NM_001267548.2 | NP_001254477.1 | 2 - 3 | 144 | 82 | |
NM_001267549.2 | NP_001254478.1 | 2 - 3 | 247 | 82 | |
NM_003224.5 | NP_003215.1 | 2 - 3 | 242 | 82 | |
NR_051954.2 | 2 - 3 | 242 | 82 | ||
NR_051955.2 | 2 - 3 | 247 | 82 | ||
NR_051956.2 | 2 - 3 | 247 | 82 | ||
NR_051957.2 | 2 - 3 | 242 | 82 | ||
NR_051958.2 | 2 - 3 | 242 | 82 | ||
XM_011528482.2 | XP_011526784.1 | 2 - 3 | 179 | 82 | |
XM_011528483.1 | XP_011526785.1 | 2 - 3 | 309 | 82 | |
XM_017027576.1 | XP_016883065.1 | 2 - 3 | 123 | 82 | |
GenBank mRNA | AK095962.1 | - | 1 - 2 | 661 | 82 |
AL522492.3 | - | 1 - 2 | 99 | 82 | |
AL533893.3 | - | 2 - 3 | 232 | 82 | |
BC021023.1 | - | 2 - 3 | 127 | 82 | |
BC092480.1 | - | 2 - 3 | 128 | 82 | |
BC093967.1 | - | 2 - 3 | 145 | 82 | |
BC143447.1 | - | 2 - 3 | 145 | 82 | |
BC143448.1 | - | 2 - 3 | 150 | 82 | |
BC143467.1 | - | 2 - 3 | 150 | 82 | |
BC143477.1 | - | 2 - 3 | 150 | 82 | |
BI524032.1 | - | 2 - 3 | 241 | 82 | |
BI819763.1 | - | 2 - 3 | 116 | 82 | |
BM549361.1 | - | - | 210 | 82 | |
BT019624.1 | - | 1 - 2 | 94 | 82 | |
BT019663.1 | - | 1 - 2 | 94 | 82 | |
BU526941.1 | - | 2 - 3 | 184 | 82 | |
BX342690.2 | - | 2 - 3 | 128 | 82 | |
BX342805.2 | - | 2 - 3 | 221 | 82 | |
BX463457.2 | - | 2 - 3 | 144 | 82 | |
CA489361.1 | - | 1 - 2 | 86 | 82 | |
CR456752.1 | - | 1 - 2 | 94 | 82 | |
X91504.1 | - | 1 - 2 | 105 | 82 |