Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
Chromosome Location
Chr.20: 63698642 - 63708013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134758.3 NP_001128230.1 6 - 7 565 107
NM_001267544.2 NP_001254473.1 6 - 7 568 107
NM_001267546.2 NP_001254475.1 5 - 6 471 107
NM_001267547.2 NP_001254476.1 6 - 7 570 107
NM_001267548.2 NP_001254477.1 6 - 7 467 107
NM_001267549.2 NP_001254478.1 6 - 7 570 107
NM_003224.5 NP_003215.1 6 - 7 565 107
NR_051954.2 6 - 7 563 107
NR_051955.2 6 - 7 568 107
NR_051956.2 6 - 7 565 107
NR_051957.2 6 - 7 560 107
XM_011528482.2 XP_011526784.1 6 - 7 502 107
XM_011528483.1 XP_011526785.1 6 - 7 632 107
XM_017027576.1 XP_016883065.1 6 - 7 446 107
GenBank mRNA AL522492.3 - 5 - 6 422 107
AL533893.3 - 6 - 7 555 107
AL833084.2 - 2 - 3 3988 107
BC021023.1 - 6 - 7 450 107
BC092480.1 - 6 - 7 451 107
BC093967.1 - 6 - 7 468 107
BC143447.1 - 6 - 7 468 107
BC143448.1 - 6 - 7 473 107
BC143467.1 - 6 - 7 473 107
BC143477.1 - 6 - 7 471 107
BI524032.1 - 6 - 7 564 107
BI819763.1 - 6 - 7 437 107
BM546222.1 - - 490 107
BT019624.1 - 5 - 6 417 107
BT019663.1 - 5 - 6 417 107
BU526941.1 - 6 - 7 502 107
BX342690.2 - 6 - 7 105
BX463457.2 - 6 - 7 467 107
CR456752.1 - 5 - 6 417 107
X91504.1 - 5 - 6 428 107

View Full Product Details