The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014747.2 | NP_055562.2 | 6 - 7 | 1047 | 95 |
XM_011542479.2 | XP_011540781.1 | 5 - 6 | 685 | 95 | |
GenBank mRNA | AY326956.1 | - | 4 - 5 | 578 | 95 |
BC003103.1 | - | 6 - 7 | 755 | 95 | |
D87074.1 | - | 6 - 7 | 1053 | 95 | |
KJ535026.1 | - | 4 - 5 | 648 | 95 |