The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005373.2 | NP_005364.1 | 1 - 2 | 119 | 56 |
GenBank mRNA | M90102.1 | - | 1 - 2 | 74 | 56 |
M90103.1 | - | 1 - 2 | 74 | 56 |