The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005373.2 | NP_005364.1 | 7 - 8 | 1211 | 64 |
XM_017001320.1 | XP_016856809.1 | 6 - 7 | 2311 | 64 | |
GenBank mRNA | DQ234353.1 | - | 7 - 8 | 1145 | 64 |
KF964490.1 | - | 1 - 2 | 68 | 64 | |
M90102.1 | - | 7 - 8 | 1166 | 64 | |
M90103.1 | - | 7 - 8 | 1166 | 64 |