Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 13 - 14 1987 94
XM_005269987.4 XP_005270044.1 13 - 14 2115 94
XM_005269988.3 XP_005270045.1 13 - 14 2046 94
XM_006717923.3 XP_006717986.1 13 - 14 2043 94
XM_006717924.3 XP_006717987.1 12 - 13 1965 94
XM_006717925.3 XP_006717988.1 13 - 14 2046 94
XM_011539978.2 XP_011538280.1 7 - 8 1319 94
XM_011539979.1 XP_011538281.1 7 - 8 1445 94
XM_011539980.2 XP_011538282.1 7 - 8 1255 94
XM_017016440.1 XP_016871929.1 7 - 8 2265 94
XM_017016441.1 XP_016871930.1 7 - 8 1223 94
XM_017016442.1 XP_016871931.1 7 - 8 1427 94
XM_017016443.1 XP_016871932.1 7 - 8 1265 94
GenBank mRNA AB033050.2 - 7 - 8 1206 94
AF357909.1 - 8 - 9 1136 94
AY235683.1 - 10 - 11 1484 94
BC007288.1 - 13 - 14 1987 94

View Full Product Details