Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 21 - 22 2997 84
XM_005269987.4 XP_005270044.1 21 - 22 3125 84
XM_005269988.3 XP_005270045.1 21 - 22 3056 84
XM_006717923.3 XP_006717986.1 21 - 22 3053 84
XM_006717924.3 XP_006717987.1 20 - 21 2975 84
XM_006717925.3 XP_006717988.1 21 - 22 3056 84
XM_011539978.2 XP_011538280.1 15 - 16 2329 84
XM_011539979.1 XP_011538281.1 15 - 16 2455 84
XM_011539980.2 XP_011538282.1 15 - 16 2265 84
XM_017016440.1 XP_016871929.1 15 - 16 3275 84
XM_017016441.1 XP_016871930.1 15 - 16 2233 84
XM_017016442.1 XP_016871931.1 15 - 16 2437 84
XM_017016443.1 XP_016871932.1 15 - 16 2275 84
GenBank mRNA AB033050.2 - 15 - 16 2216 84
AY235683.1 - 18 - 19 2494 84
BE082814.1 - 2 - 3 94 84

View Full Product Details