Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 3 - 4 446 127
XM_005269987.4 XP_005270044.1 3 - 4 556 127
XM_005269988.3 XP_005270045.1 3 - 4 487 127
XM_006717925.3 XP_006717988.1 3 - 4 487 127
GenBank mRNA BC007288.1 - 3 - 4 446 127
BX644960.1 - 1 - 2 58 127

View Full Product Details