Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 6 - 7 747 73
XM_005269987.4 XP_005270044.1 6 - 7 857 73
XM_005269988.3 XP_005270045.1 6 - 7 788 73
XM_006717923.3 XP_006717986.1 6 - 7 785 73
XM_006717924.3 XP_006717987.1 5 - 6 707 73
XM_006717925.3 XP_006717988.1 6 - 7 788 73
GenBank mRNA AK025812.1 - 4 - 5 329 73
AY235683.1 - 3 - 4 244 73
BC007288.1 - 6 - 7 747 73

View Full Product Details