Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 7 - 8 852 63
XM_005269987.4 XP_005270044.1 7 - 8 962 63
XM_005269988.3 XP_005270045.1 7 - 8 893 63
XM_006717923.3 XP_006717986.1 7 - 8 890 63
XM_006717924.3 XP_006717987.1 6 - 7 812 63
XM_006717925.3 XP_006717988.1 7 - 8 893 63
GenBank mRNA AY235683.1 - 4 - 5 349 63
BC007288.1 - 7 - 8 852 63
BC117687.2 - 3 - 4 318 63

View Full Product Details