The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145546.1 | NP_001139018.1 | 4 - 5 | 333 | 117 |
NM_033106.3 | NP_149097.1 | 5 - 6 | 382 | 117 | |
GenBank mRNA | AF188492.1 | - | 5 - 6 | 382 | 117 |
AF188493.1 | - | 5 - 6 | 382 | 117 |