Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
Chromosome Location
Chr.2: 120797291 - 120992653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005270.4 NP_005261.2 7 - 8 1213 90
XM_011510969.2 XP_011509271.2 7 - 8 1845 90
XM_011510970.2 XP_011509272.1 7 - 8 2856 90
XM_011510971.2 XP_011509273.1 7 - 8 2106 90
XM_011510972.2 XP_011509274.2 7 - 8 1243 90
XM_011510973.2 XP_011509275.1 6 - 7 994 90
GenBank mRNA DQ086814.1 - 7 - 8 1183 90

View Full Product Details