The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288607.1 | NP_001275536.1 | 13 - 14 | 2215 | 71 |
NM_001288608.1 | NP_001275537.1 | 13 - 14 | 2401 | 71 | |
NM_019032.5 | NP_061905.2 | 12 - 13 | 2332 | 71 | |
NM_025008.4 | NP_079284.2 | 12 - 13 | 2332 | 71 | |
XM_011509644.2 | XP_011507946.1 | 12 - 13 | 2363 | 71 | |
XM_011509645.2 | XP_011507947.1 | 11 - 12 | 2291 | 71 | |
XM_011509648.2 | XP_011507950.1 | 12 - 13 | 2317 | 71 | |
XM_011509649.2 | XP_011507951.1 | 12 - 13 | 2371 | 71 | |
XM_011509650.2 | XP_011507952.1 | 12 - 13 | 2371 | 71 | |
XM_011509651.2 | XP_011507953.1 | 6 - 7 | 1113 | 71 | |
XM_011509652.1 | XP_011507954.1 | 6 - 7 | 819 | 71 | |
XM_017001506.1 | XP_016856995.1 | 13 - 14 | 2772 | 71 | |
XM_017001507.1 | XP_016856996.1 | 4 - 5 | 554 | 71 | |
GenBank mRNA | AF217974.1 | - | 1 - 2 | 134 | 71 |
AF370411.1 | - | 6 - 7 | 868 | 71 | |
AK092509.1 | - | 3 - 4 | 452 | 71 | |
AK293037.1 | - | 2 - 3 | 264 | 71 | |
AY358122.1 | - | 11 - 12 | 2146 | 71 | |
BC027478.1 | - | 5 - 6 | 771 | 71 | |
BC071852.1 | - | 4 - 5 | 628 | 71 | |
BC094811.1 | - | 4 - 5 | 628 | 71 | |
BC140800.1 | - | 12 - 13 | 2253 | 71 | |
BC144581.1 | - | 12 - 13 | 2067 | 71 |