Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
Chromosome Location
Chr.1: 150549369 - 150560937 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288607.1 NP_001275536.1 16 - 17 2729 93
NM_001288608.1 NP_001275537.1 16 - 17 2915 93
NM_019032.5 NP_061905.2 15 - 16 2846 93
XM_011509644.2 XP_011507946.1 15 - 16 2877 93
XM_011509645.2 XP_011507947.1 14 - 15 2805 93
XM_011509648.2 XP_011507950.1 15 - 16 2831 93
XM_011509651.2 XP_011507953.1 9 - 10 1627 93
XM_011509652.1 XP_011507954.1 9 - 10 1333 93
XM_017001506.1 XP_016856995.1 16 - 17 3286 93
XM_017001507.1 XP_016856996.1 7 - 8 1068 93
GenBank mRNA AF217974.1 - 4 - 5 647 93
AK092509.1 - 6 - 7 965 93
AK293037.1 - 5 - 6 778 93
BC027478.1 - 8 - 9 1285 93
BC071852.1 - 7 - 8 1142 93
BC094811.1 - 7 - 8 1142 93
BC140800.1 - 15 - 16 2767 93
BC144581.1 - 15 - 16 2581 93
BX647294.1 - 2 - 3 408 93

View Full Product Details