The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006255.4 | NP_006246.2 | 14 - 14 | 3710 | 130 |
XM_011536954.2 | XP_011535256.1 | 14 - 14 | 2893 | 130 | |
XM_011536955.1 | XP_011535257.1 | 14 - 14 | 2826 | 130 | |
XM_017021458.1 | XP_016876947.1 | 14 - 14 | 3044 | 130 | |
GenBank mRNA | AK093241.1 | - | 3 - 3 | 2603 | 130 |
AW008462.1 | - | 1 - 1 | 130 | ||
AX775931.1 | - | 14 - 14 | 2786 | 130 | |
AX775933.1 | - | 14 - 14 | 3439 | 130 | |
BC001000.2 | - | 1 - 1 | 875 | 130 | |
BC037268.1 | - | 14 - 14 | 3439 | 130 | |
CA447784.1 | - | 1 - 1 | 130 |