Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BIN1
- Entrez Gene ID
-
274
- Gene Name
- bridging integrator 1
- Gene Aliases
-
AMPH2, AMPHL, SH3P9
- Chromosome Location
-
Chr.2: 127048023 - 127107400 on Build GRCh38
View Full Product Details