Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN1A
Entrez Gene ID
1026
Gene Name
cyclin dependent kinase inhibitor 1A
Gene Aliases
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
Chromosome Location
Chr.6: 36676460 - 36687339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291549.1 NP_001278478.1 2 - 3 274 86
GenBank mRNA AK298901.1 - 1 - 2 131 86
BU193165.1 - 2 - 3 274 86

View Full Product Details