The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006910.4 | NP_008841.2 | 9 - 10 | 1993 | 134 |
NM_018703.3 | NP_061173.1 | 9 - 10 | 1993 | 134 | |
GenBank mRNA | AB112074.1 | - | 9 - 10 | 997 | 134 |
AB112075.1 | - | 9 - 10 | 997 | 134 | |
AF352051.1 | - | 4 - 5 | 516 | 134 | |
AK303040.1 | - | 7 - 8 | 891 | 134 | |
AY072922.1 | - | 5 - 6 | 1742 | 134 | |
BC063524.1 | - | 9 - 10 | 1060 | 134 | |
BX648661.1 | - | 9 - 10 | 933 | 134 | |
X85133.1 | - | 4 - 5 | 516 | 134 |