The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018436.3 | NP_060906.3 | 4 - 5 | 335 | 105 |
XM_011510369.2 | XP_011508671.1 | 3 - 4 | 229 | 105 | |
XM_017004495.1 | XP_016859984.1 | 5 - 6 | 1414 | 105 | |
XM_017004496.1 | XP_016859985.1 | 2 - 3 | 1375 | 105 | |
GenBank mRNA | AF395820.1 | - | 2 - 3 | 131 | 105 |
AK302299.1 | - | 4 - 5 | 335 | 105 | |
BC029652.1 | - | 4 - 5 | 311 | 105 |