The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015474.3 | NP_056289.2 | 11 - 12 | 1466 | 88 |
XM_005260384.3 | XP_005260441.1 | 11 - 12 | 1439 | 88 | |
XM_011528761.2 | XP_011527063.1 | 11 - 12 | 1439 | 88 | |
GenBank mRNA | AB013847.1 | - | 11 - 12 | 1266 | 88 |
AB208944.1 | - | 11 - 12 | 1291 | 88 | |
AF228421.1 | - | 11 - 12 | 1419 | 88 | |
AK027811.1 | - | 11 - 12 | 1439 | 88 | |
AK304187.1 | - | 11 - 12 | 1223 | 88 | |
AK315169.1 | - | 11 - 12 | 1399 | 88 | |
AL050267.1 | - | 11 - 12 | 1341 | 88 | |
BC036450.2 | - | 11 - 12 | 1294 | 88 |