Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
Chromosome Location
Chr.7: 100679507 - 100694282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022574.4 NP_072096.2 11 - 12 2300 56
XM_005250532.1 XP_005250589.1 13 - 14 2081 56
XM_011516472.1 XP_011514774.1 13 - 14 2081 56
XM_011516477.1 XP_011514779.1 13 - 14 2081 56
XM_017012526.1 XP_016868015.1 13 - 14 2081 56
XM_017012527.1 XP_016868016.1 15 - 16 1979 56
XM_017012528.1 XP_016868017.1 14 - 15 1591 56
XM_017012529.1 XP_016868018.1 14 - 15 1598 56
XM_017012530.1 XP_016868019.1 14 - 15 1852 56
XM_017012531.1 XP_016868020.1 9 - 10 1104 56
GenBank mRNA AY176044.1 - 11 - 12 2501 56
BC129991.1 - 5 - 6 448 56

View Full Product Details