The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000809.3 | NP_000800.2 | 2 - 3 | 1187 | 96 |
NM_001204266.1 | NP_001191195.1 | 2 - 3 | 217 | 96 | |
NM_001204267.1 | NP_001191196.1 | 2 - 3 | 217 | 96 | |
GenBank mRNA | AK090780.1 | - | 2 - 3 | 1187 | 96 |
BC035055.1 | - | 2 - 3 | 316 | 96 | |
BI599812.1 | - | 2 - 3 | 347 | 96 | |
CD013892.1 | - | 2 - 3 | 221 | 96 | |
CD013893.1 | - | 2 - 3 | 221 | 96 | |
KJ534840.1 | - | 2 - 3 | 204 | 96 | |
KJ535037.1 | - | 2 - 3 | 204 | 96 | |
U30461.1 | - | 2 - 3 | 218 | 96 |