The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000809.3 | NP_000800.2 | 4 - 5 | 1479 | 153 |
NM_001204266.1 | NP_001191195.1 | 4 - 5 | 509 | 153 | |
NM_001204267.1 | NP_001191196.1 | 4 - 5 | 509 | 153 | |
GenBank mRNA | AK090780.1 | - | 4 - 5 | 1479 | 153 |
BC035055.1 | - | 4 - 5 | 608 | 153 | |
CD013892.1 | - | 4 - 5 | 513 | 153 | |
CD013893.1 | - | 4 - 5 | 513 | 153 | |
KJ534840.1 | - | 4 - 5 | 496 | 153 | |
U30461.1 | - | 4 - 5 | 510 | 153 |