Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK5
Entrez Gene ID
8645
Gene Name
potassium two pore domain channel subfamily K member 5
Gene Aliases
K2p5.1, KCNK5b, TASK-2, TASK2
Chromosome Location
Chr.6: 39188971 - 39229475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003740.3 NP_003731.1 2 - 3 662 76
XM_005249456.1 XP_005249513.1 2 - 3 395 76
GenBank mRNA AF084830.1 - 2 - 3 637 76
AK314298.1 - 2 - 3 395 76
BC060793.1 - 2 - 3 662 76
BC069573.1 - 2 - 3 369 76
EU978936.1 - 2 - 3 448 76

View Full Product Details