Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14orf79
Entrez Gene ID
122616
Gene Name
chromosome 14 open reading frame 79
Gene Aliases
-
Chromosome Location
Chr.14: 104985801 - 105000310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174891.3 NP_777551.2 3 - 4 847 105
XM_005267317.3 XP_005267374.1 3 - 4 1325 105
XM_005267318.3 XP_005267375.1 3 - 4 1325 105
GenBank mRNA AF038188.1 - 3 - 4 411 105
AK057249.1 - 4 - 5 923 105
BC003539.1 - 2 - 3 159 105
BC011537.1 - 2 - 3 677 105
BC025175.1 - 3 - 4 358 105
BC137495.1 - 3 - 4 790 105
BC144671.1 - 3 - 4 776 105

View Full Product Details