Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RLF
Entrez Gene ID
6018
Gene Name
rearranged L-myc fusion
Gene Aliases
ZN-15L, ZNF292L
Chromosome Location
Chr.1: 40161369 - 40240921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012421.3 NP_036553.2 4 - 5 637 107
XM_017002017.1 XP_016857506.1 4 - 5 617 107
XM_017002018.1 XP_016857507.1 4 - 5 617 107
GenBank mRNA AK307662.1 - 4 - 5 621 107
BC113666.1 - 4 - 5 637 107
U22377.1 - 4 - 5 618 107

View Full Product Details