The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292045.1 | NP_001278974.1 | 1 - 2 | 152 | |
NM_001292046.1 | NP_001278975.1 | 1 - 2 | 152 | ||
NM_006681.3 | NP_006672.1 | 1 - 2 | 152 | ||
GenBank mRNA | BC012908.1 | - | 1 - 2 | 152 | |
BF185217.1 | - | 1 - 2 | 152 | ||
BI603117.1 | - | 1 - 2 | 222 | 153 | |
BQ049320.1 | - | 1 - 2 | 152 | ||
X76029.1 | - | 1 - 2 | 152 |