The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005028.4 | NP_005019.2 | 1 - 2 | 392 | 81 |
XM_006717450.2 | XP_006717513.1 | 1 - 2 | 387 | 81 | |
GenBank mRNA | BC018034.1 | - | 1 - 2 | 373 | 81 |
DB504104.1 | - | 1 - 2 | 393 | 81 | |
S78798.1 | - | 1 - 2 | 458 | 81 | |
U14957.2 | - | 1 - 2 | 166 | 81 |