The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005028.4 | NP_005019.2 | 8 - 9 | 1285 | 121 |
XM_006717450.2 | XP_006717513.1 | 7 - 8 | 1127 | 121 | |
XM_011519496.1 | XP_011517798.1 | 8 - 9 | 1061 | 121 | |
XM_017016330.1 | XP_016871819.1 | 8 - 9 | 1119 | 121 | |
XM_017016331.1 | XP_016871820.1 | 8 - 9 | 1214 | 121 | |
XM_017016332.1 | XP_016871821.1 | 7 - 8 | 927 | 121 | |
GenBank mRNA | AK294817.1 | - | 8 - 9 | 1066 | 121 |
AK294867.1 | - | 6 - 7 | 672 | 121 | |
BC018034.1 | - | 8 - 9 | 1266 | 121 | |
S78798.1 | - | 8 - 9 | 1351 | 121 | |
U14957.2 | - | 8 - 9 | 1059 | 121 |