The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300948.1 | NP_001287877.1 | 10 - 11 | 1881 | 99 |
NM_031490.3 | NP_113678.2 | 11 - 12 | 2013 | 99 | |
XM_017023755.1 | XP_016879244.1 | 11 - 12 | 1880 | 99 | |
XM_017023756.1 | XP_016879245.1 | 10 - 11 | 1746 | 99 | |
GenBank mRNA | AJ548761.1 | - | 11 - 12 | 1880 | 99 |
AK027544.1 | - | 11 - 12 | 1846 | 99 | |
AK027666.1 | - | 7 - 8 | 1048 | 99 | |
AK074775.1 | - | 11 - 12 | 1878 | 99 | |
AK127125.1 | - | 11 - 12 | 1645 | 99 | |
AK172790.1 | - | 2 - 3 | 168 | 99 | |
AK294779.1 | - | 10 - 11 | 1693 | 99 | |
AL834201.1 | - | 10 - 11 | 1379 | 99 | |
BC093910.1 | - | 11 - 12 | 1847 | 99 | |
BC093912.1 | - | 11 - 12 | 1847 | 99 | |
BC110434.1 | - | 7 - 8 | 1029 | 99 | |
BC143246.1 | - | 10 - 11 | 1715 | 99 |