The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010989.2 | NP_001010989.1 | 7 - 8 | 1166 | 126 |
NM_001272103.1 | NP_001259032.1 | 7 - 8 | 1012 | 126 | |
NM_014685.3 | NP_055500.1 | 7 - 8 | 1169 | 126 | |
GenBank mRNA | AB034989.1 | - | 7 - 8 | 1112 | 126 |
AF055001.1 | - | 7 - 8 | 1109 | 126 | |
AF055003.1 | - | 7 - 8 | 909 | 126 | |
AF217990.1 | - | 4 - 5 | 1450 | 126 | |
AK223233.1 | - | 7 - 8 | 1112 | 126 | |
AK291565.1 | - | 7 - 8 | 1112 | 126 | |
AK292088.1 | - | 7 - 8 | 1112 | 126 | |
AK302167.1 | - | 6 - 7 | 1073 | 126 | |
BC000086.1 | - | 7 - 8 | 1104 | 126 | |
BC008320.1 | - | 7 - 8 | 1103 | 126 | |
BC009739.1 | - | 5 - 6 | 635 | 126 | |
BC032673.1 | - | 7 - 8 | 1111 | 126 | |
CR457116.1 | - | 7 - 8 | 1014 | 126 | |
D14695.1 | - | 7 - 8 | 1107 | 126 | |
DQ837586.1 | - | 7 - 8 | 954 | 126 |