The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300791.1 | NP_001287720.1 | 1 - 2 | 160 | 137 |
NM_001300792.1 | NP_001287721.1 | 1 - 2 | 160 | 137 | |
NM_007054.6 | NP_008985.3 | 1 - 2 | 160 | 137 | |
XM_006714526.3 | XP_006714589.1 | 1 - 2 | 215 | 137 | |
XM_017008995.1 | XP_016864484.1 | 1 - 2 | 215 | 137 | |
XM_017008996.1 | XP_016864485.1 | 1 - 2 | 215 | 137 | |
GenBank mRNA | AB209780.1 | - | 1 - 2 | 108 | 137 |
AK295089.1 | - | 1 - 2 | 118 | 137 | |
AK313359.1 | - | 1 - 2 | 128 | 137 | |
BC020890.1 | - | 1 - 2 | 126 | 137 | |
BC032599.1 | - | 1 - 2 | 143 | 137 | |
BC039592.1 | - | 1 - 2 | 117 | 137 | |
BC045542.1 | - | 1 - 2 | 154 | 137 | |
BI561509.1 | - | 1 - 2 | 160 | 137 | |
HY154618.1 | - | 1 - 2 | 160 | 137 |