The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032313.3 | NP_115689.1 | 3 - 4 | 1578 | 109 |
GenBank mRNA | AK074953.1 | - | 3 - 4 | 1506 | 109 |
AK098215.1 | - | 4 - 5 | 1455 | 109 | |
BC004894.2 | - | 3 - 4 | 1578 | 109 |