The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003213.3 | NP_003204.2 | 11 - 12 | 1329 | 125 |
NM_201441.2 | NP_958849.1 | 10 - 11 | 1200 | 125 | |
NM_201443.2 | NP_958851.1 | 9 - 10 | 1270 | 125 | |
GenBank mRNA | AK222947.1 | - | 11 - 12 | 1216 | 125 |
AY101179.1 | - | 10 - 11 | 1182 | 125 | |
BC015497.1 | - | 11 - 12 | 1227 | 125 | |
BX640921.1 | - | 9 - 10 | 936 | 125 | |
KU178354.1 | - | 8 - 9 | 818 | 125 | |
KU178355.1 | - | 5 - 6 | 589 | 125 | |
KU178356.1 | - | 4 - 5 | 449 | 125 | |
U63824.1 | - | 11 - 12 | 1200 | 125 | |
X94438.1 | - | 9 - 10 | 1016 | 125 |