The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003213.3 | NP_003204.2 | 9 - 10 | 1018 | 77 |
NM_201441.2 | NP_958849.1 | 8 - 9 | 889 | 77 | |
NM_201443.2 | NP_958851.1 | 7 - 8 | 959 | 77 | |
GenBank mRNA | AK222947.1 | - | 9 - 10 | 905 | 77 |
AY101179.1 | - | 8 - 9 | 871 | 77 | |
BC015497.1 | - | 9 - 10 | 916 | 77 | |
BX640921.1 | - | 7 - 8 | 625 | 77 | |
KU178354.1 | - | 6 - 7 | 507 | 77 | |
KU178355.1 | - | 3 - 4 | 278 | 77 | |
U63824.1 | - | 9 - 10 | 889 | 77 | |
X94438.1 | - | 7 - 8 | 705 | 77 |