The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001496.3 | NP_001487.2 | 5 - 6 | 1033 | 75 |
GenBank mRNA | AF051767.1 | - | 5 - 6 | 893 | 75 |
AK297693.1 | - | 4 - 5 | 894 | 75 | |
AK314022.1 | - | 5 - 6 | 934 | 75 | |
AY358997.1 | - | 5 - 6 | 930 | 75 | |
AY359037.1 | - | 4 - 5 | 941 | 75 | |
BC037951.1 | - | 5 - 6 | 918 | 75 | |
FJ200487.1 | - | 4 - 5 | 800 | 75 |