The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015080.3 | NP_055895.1 | 12 - 13 | 2879 | 87 |
XM_005274400.3 | XP_005274457.1 | 11 - 12 | 2411 | 87 | |
XM_005274401.3 | XP_005274458.1 | 11 - 12 | 2411 | 87 | |
XM_005274402.3 | XP_005274459.1 | 10 - 11 | 2387 | 87 | |
XM_017018568.1 | XP_016874057.1 | 11 - 12 | 2432 | 87 | |
XM_017018571.1 | XP_016874060.1 | 11 - 12 | 2432 | 87 | |
XM_017018577.1 | XP_016874066.1 | 4 - 5 | 777 | 87 | |
XM_017018578.1 | XP_016874067.1 | 11 - 12 | 2432 | 87 | |
GenBank mRNA | AB035266.1 | - | 11 - 12 | 2531 | 87 |
AB177862.1 | - | 8 - 9 | 1583 | 87 |