Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 8 - 9 1823 78
NM_138732.2 NP_620060.1 7 - 8 1730 78
XM_005274400.3 XP_005274457.1 7 - 8 1355 78
XM_005274401.3 XP_005274458.1 7 - 8 1355 78
XM_005274402.3 XP_005274459.1 6 - 7 1331 78
XM_011545370.1 XP_011543672.1 7 - 8 1376 78
XM_011545371.1 XP_011543673.1 7 - 8 1373 78
XM_011545373.1 XP_011543675.1 7 - 8 1355 78
XM_011545375.1 XP_011543677.1 6 - 7 1331 78
XM_017018563.1 XP_016874052.1 6 - 7 1346 78
XM_017018564.1 XP_016874053.1 6 - 7 1343 78
XM_017018565.1 XP_016874054.1 7 - 8 1376 78
XM_017018566.1 XP_016874055.1 6 - 7 1304 78
XM_017018567.1 XP_016874056.1 7 - 8 1376 78
XM_017018568.1 XP_016874057.1 7 - 8 1376 78
XM_017018569.1 XP_016874058.1 5 - 6 1274 78
XM_017018570.1 XP_016874059.1 7 - 8 1376 78
XM_017018571.1 XP_016874060.1 7 - 8 1376 78
XM_017018572.1 XP_016874061.1 7 - 8 671 78
XM_017018573.1 XP_016874062.1 7 - 8 1376 78
XM_017018574.1 XP_016874063.1 6 - 7 641 78
XM_017018575.1 XP_016874064.1 6 - 7 638 78
XM_017018576.1 XP_016874065.1 2 - 3 286 78
XM_017018578.1 XP_016874067.1 7 - 8 1376 78
GenBank mRNA AB023138.2 - 8 - 9 1707 78
AB035266.1 - 7 - 8 1475 78
AB177862.1 - 4 - 5 527 78
BC150275.1 - 8 - 9 1707 78

View Full Product Details