Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APBA2
Entrez Gene ID
321
Gene Name
amyloid beta precursor protein binding family A member 2
Gene Aliases
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
Chromosome Location
Chr.15: 28885479 - 29118315 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005503.3 NP_005494.2 7 - 8 1461 96
XM_011521488.2 XP_011519790.1 8 - 9 1597 96
XM_011521489.2 XP_011519791.1 9 - 10 2448 96
XM_011521490.2 XP_011519792.1 9 - 10 2417 96
XM_011521491.1 XP_011519793.1 7 - 8 1476 96
XM_011521492.2 XP_011519794.1 7 - 8 1726 96
XM_011521494.2 XP_011519796.1 8 - 9 2258 96
XM_011521495.1 XP_011519797.1 5 - 6 745 96
XM_017022101.1 XP_016877590.1 8 - 9 2258 96
XM_017022102.1 XP_016877591.1 9 - 10 2348 96
XM_017022103.1 XP_016877592.1 9 - 10 1667 96
XM_017022104.1 XP_016877593.1 8 - 9 1690 96
XM_017022105.1 XP_016877594.1 8 - 9 1643 96
XM_017022106.1 XP_016877595.1 8 - 9 2254 96
XM_017022107.1 XP_016877596.1 8 - 9 1723 96
XM_017022108.1 XP_016877597.1 8 - 9 2223 96
XM_017022109.1 XP_016877598.1 9 - 10 2344 96
XM_017022110.1 XP_016877599.1 9 - 10 2444 96
XM_017022111.1 XP_016877600.1 7 - 8 1587 96
XM_017022112.1 XP_016877601.1 8 - 9 1640 96
GenBank mRNA AB014719.1 - 5 - 6 1275 96
AB209281.1 - 6 - 7 1332 96
AF047348.1 - 7 - 8 1452 96

View Full Product Details