The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000227.4 | NP_000218.3 | 3 - 4 | 424 | 91 |
NM_001127717.2 | NP_001121189.2 | 40 - 41 | 5467 | 91 | |
NM_001127718.2 | NP_001121190.2 | 3 - 4 | 424 | 91 | |
NM_198129.2 | NP_937762.2 | 40 - 41 | 5467 | 91 | |
XM_011525978.2 | XP_011524280.1 | 40 - 41 | 5494 | 91 | |
XM_011525979.2 | XP_011524281.1 | 40 - 41 | 5485 | 91 | |
XM_011525980.2 | XP_011524282.1 | 40 - 41 | 5476 | 91 | |
XM_011525981.2 | XP_011524283.1 | 39 - 40 | 5362 | 91 | |
XM_011525982.2 | XP_011524284.1 | 40 - 41 | 5494 | 91 | |
XM_017025743.1 | XP_016881232.1 | 23 - 24 | 3178 | 91 | |
XM_017025744.1 | XP_016881233.1 | 13 - 14 | 1859 | 91 | |
GenBank mRNA | AB107369.1 | - | 40 - 41 | 5226 | 91 |
AY327114.1 | - | 3 - 4 | 399 | 91 | |
AY327115.1 | - | 40 - 41 | 5226 | 91 | |
AY327116.1 | - | 40 - 41 | 5226 | 91 | |
DA862923.1 | - | 3 - 4 | 424 | 91 | |
L34155.1 | - | 3 - 4 | 366 | 91 | |
X84392.1 | - | 3 - 4 | 401 | 91 | |
X84900.1 | - | 5 - 6 | 645 | 91 |