The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004690.3 | NP_004681.1 | 6 - 7 | 3318 | 103 |
NR_073033.1 | 7 - 8 | 3224 | 103 | ||
XM_011536252.2 | XP_011534554.1 | 6 - 7 | 3160 | 103 | |
XM_017011474.1 | XP_016866963.1 | 6 - 7 | 3160 | 103 | |
XM_017011475.1 | XP_016866964.1 | 6 - 7 | 3080 | 103 | |
XM_017011476.1 | XP_016866965.1 | 7 - 8 | 3228 | 103 | |
XM_017011477.1 | XP_016866966.1 | 6 - 7 | 3160 | 103 | |
XM_017011478.1 | XP_016866967.1 | 7 - 8 | 3237 | 103 | |
XM_017011479.1 | XP_016866968.1 | 6 - 7 | 3160 | 103 | |
XM_017011480.1 | XP_016866969.1 | 5 - 6 | 2857 | 103 | |
XM_017011481.1 | XP_016866970.1 | 5 - 6 | 3002 | 103 | |
GenBank mRNA | AF104413.1 | - | 6 - 7 | 3318 | 103 |
AF164041.1 | - | 5 - 6 | 2802 | 103 | |
AK310422.1 | - | 7 - 8 | 3224 | 103 |