Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHB2
Entrez Gene ID
56133
Gene Name
protocadherin beta 2
Gene Aliases
PCDH-BETA2
Chromosome Location
Chr.5: 141094607 - 141097380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018936.3 NP_061759.1 1 - 1 1316 139
GenBank mRNA AB209745.1 - 1 - 1 1092 139
AF152495.1 - 1 - 1 1132 139
AF217756.1 - 1 - 1 1132 139
AK027526.1 - 1 - 1 1287 139
AK314642.1 - 1 - 1 1270 139
BC098347.1 - 1 - 1 1179 139
BC098575.1 - 1 - 1 1280 139

View Full Product Details