The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100112.1 | NP_001093582.1 | 3 - 4 | 334 | 108 |
NM_017534.5 | NP_060004.3 | 3 - 4 | 334 | 108 | |
GenBank mRNA | AF111784.1 | - | 1 - 2 | 206 | 108 |
AK300702.1 | - | 3 - 4 | 292 | 108 | |
AK308102.1 | - | 3 - 4 | 292 | 108 | |
BC093082.1 | - | 2 - 3 | 279 | 108 | |
BC126409.1 | - | 2 - 3 | 277 | 108 | |
BX510904.2 | - | 3 - 4 | 297 | 108 | |
DA765261.1 | - | 3 - 4 | 334 | 107 | |
DC379233.1 | - | 3 - 4 | 334 | 108 |