The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015166.3 | NP_055981.1 | 8 - 9 | 1349 | 63 |
NM_139202.2 | NP_631941.1 | 8 - 9 | 988 | 63 | |
XM_011530678.1 | XP_011528980.1 | 8 - 9 | 1110 | 63 | |
XM_017028671.1 | XP_016884160.1 | 7 - 8 | 777 | 63 | |
GenBank mRNA | AF319633.1 | - | 8 - 9 | 911 | 63 |
AK124264.1 | - | 8 - 9 | 1350 | 63 | |
AK294048.1 | - | 6 - 7 | 739 | 63 | |
AK295106.1 | - | 7 - 8 | 610 | 63 | |
AK297341.1 | - | 7 - 8 | 709 | 63 | |
AK299841.1 | - | 7 - 8 | 757 | 63 | |
BC028425.2 | - | 8 - 9 | 948 | 63 | |
BC070042.1 | - | 8 - 9 | 946 | 63 | |
CR456460.1 | - | 7 - 8 | 767 | 63 | |
D25217.3 | - | 8 - 9 | 1025 | 63 |