The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084.4 | NP_001075.1 | 2 - 3 | 653 | 84 |
GenBank mRNA | AF046889.1 | - | 2 - 3 | 413 | 84 |
AF068229.1 | - | 2 - 3 | 258 | 84 | |
AK075338.1 | - | 2 - 3 | 501 | 84 | |
AK297559.1 | - | 2 - 3 | 234 | 84 | |
AK298537.1 | - | 3 - 4 | 340 | 84 | |
AK312743.1 | - | 2 - 3 | 411 | 84 | |
AY220458.1 | - | 2 - 3 | 261 | 84 | |
BC011674.2 | - | 2 - 3 | 510 | 84 |